Learn More
Strem, An Ascensus Company CAS# 10361-79-2. 5g. Praseodymium(III) chloride, anhydrous (99.9%-Pr) (REO). MFCD00011176
SDP
Catalog No. 5090115913
Encompass
CAS# 10361-79-2. 5g. Praseodymium(III) chloride, anhydrous (99.9%-Pr) (REO). MFCD00011176. Molecular Weight: 247.24. Molecular Formula: PrCl3. Color/form: pale green to blue pwdr. Strem# 93-5905. http://www.strem.com/catalog/v/93-5905/
Catalog No. 50-901-15913 Supplier Strem, An Ascensus Company Supplier No. 9359055G
CAS# 10361-79-2. 5g. Praseodymium(III) chloride, anhydrous (99.9%-Pr) (REO). MFCD00011176. Molecular Weight: 247.24. Molecular Formula: PrCl3. Color/form: pale green to blue pwdr. Strem# 93-5905. http://www.strem.com/catalog/v/93-5905/