Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) iodide, ultra dry, 99.9+% (REO)

Catalog No. AA3938506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3938506 5 g
AA3938503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3938506 Supplier Thermo Scientific Chemicals Supplier No. 03938506
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13813-23-5 | I3Pr | 521.621 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-23-5
Molecular Formula I3Pr
Molecular Weight (g/mol) 521.621
MDL Number MFCD00049513
InChI Key PVEVRIVGNKNWML-UHFFFAOYSA-K
Synonym praseodymium triiodide, praseodymium iodide pri3, pri3, prazodymium iodide, praseodymium iii iodide, ultra dry
PubChem CID 83744
IUPAC Name triiodopraseodymium
SMILES I[Pr](I)I

Specifications

Melting Point 737°C
Physical Form Crystals or Needles
Quantity 5 g
Linear Formula PrI3
Solubility Information Soluble in water.
Formula Weight 521.62
Percent Purity ≥99.9%
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Assay (REO)
Chemical Name or Material Praseodymium(III) iodide, Ultra dry
Show More Show Less
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 237-466-1
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.