Learn More
Strem, An Ascensus Company CAS# 12037-29-5. 250g. Praseodymium(III,IV) oxide (99.9%-Pr) (REO). MFCD00011178
SDP
Catalog No. 5090115926
Encompass
CAS# 12037-29-5. 250g. Praseodymium(III,IV) oxide (99.9%-Pr) (REO). MFCD00011178. Molecular Weight: 1021.43. Molecular Formula: Pr6O11. Color/form: black pwdr. Strem# 93-5911. http://www.strem.com/catalog/v/93-5911/
Catalog No. 50-901-15926 Supplier Strem, An Ascensus Company Supplier No. 935911250G
CAS# 12037-29-5. 250g. Praseodymium(III,IV) oxide (99.9%-Pr) (REO). MFCD00011178. Molecular Weight: 1021.43. Molecular Formula: Pr6O11. Color/form: black pwdr. Strem# 93-5911. http://www.strem.com/catalog/v/93-5911/