Learn More
Apexbio Technology LLC Prasugrel 150322-43-3 100mg
SDP
Catalog No. 502240787
Encompass_Preferred
Prasugrel (CAS 150322-43-3) is a small molecule inhibitor belonging to the thienopyridine chemical class that targets the P2Y12 subtype of adenosine diphosphate (ADP) receptors on platelets Through irreversible antagonism of these receptors prasugrel prevents ADP-mediated activation and aggregation of platelets This mode of action makes it a valuable tool for investigating platelet function thrombosis and the modulation of hemostatic pathways in preclinical and translational research models Prasugrel is frequently utilized in studies exploring the pharmacological regulation of platelet reactivity and ADP receptor signaling
This item is not returnable. View return policy
Catalog No. 50-224-0787 Supplier Apexbio Technology LLC Supplier No. A1607100
Prasugrel (CAS 150322-43-3) is a small molecule inhibitor belonging to the thienopyridine chemical class that targets the P2Y12 subtype of adenosine diphosphate (ADP) receptors on platelets Through irreversible antagonism of these receptors prasugrel prevents ADP-mediated activation and aggregation of platelets This mode of action makes it a valuable tool for investigating platelet function thrombosis and the modulation of hemostatic pathways in preclinical and translational research models Prasugrel is frequently utilized in studies exploring the pharmacological regulation of platelet reactivity and ADP receptor signaling