Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prasugrel-d3 | 1127253-02-4 | MFCD09954140 | 98.6% | 376.46 | C20H17D3FNO3S | 10 MG
SDP

Catalog No. 5000286196
Encompass_Preferred

Prasugrel-d3 is the deuterium-labeled analog of prasugrel used as an internal standard and tracer in quantitative analytical methods. It is supplied as a purified white to off-white solid in small-mass vials suitable for research, method development, and calibration workflows for mass spectrometry and NMR analyses.

  • Deuterium-labeled internal standard for quantitative LC-MS, GC-MS, and NMR.
  • High chemical purity (98.58%).
  • Molecular weight 376.46 and formula C20H17D3FNO3S.
  • Supplied in multiple small pack sizes including 10 MG.
  • Appropriate for tracer, calibration, and method validation applications.

Catalog No. 50-002-86196 Supplier Medchemexpress LLC Supplier No. HY15284S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prasugrel-d3 is the deuterium-labeled analog of prasugrel used as an internal standard and tracer in quantitative analytical methods. It is supplied as a purified white to off-white solid in small-mass vials suitable for research, method development, and calibration workflows for mass spectrometry and NMR analyses.

  • Deuterium-labeled internal standard for quantitative LC-MS, GC-MS, and NMR.
  • High chemical purity (98.58%).
  • Molecular weight 376.46 and formula C20H17D3FNO3S.
  • Supplied in multiple small pack sizes including 10 MG.
  • Appropriate for tracer, calibration, and method validation applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.