Learn More
Selleck Chemical LLC Prasugrel Hydrochloride S4637-100mg
SDP
Catalog No. 503111470
Encompass_Preferred
Prasugrel hydrochloride (Prasugrel HCl LY640315 PCR 4099 Hydrochloride) is a piperazine derivative and pletelet aggregation inhibitor that is used to prevent thrombosis in patients with acute coronary syndrome
Catalog No. 50-311-1470 Supplier Selleck Chemical LLC Supplier No. S4637100MG
Prasugrel hydrochloride (Prasugrel HCl LY640315 PCR 4099 Hydrochloride) is a piperazine derivative and pletelet aggregation inhibitor that is used to prevent thrombosis in patients with acute coronary syndrome