Learn More
Medchemexpress LLC HY-15284A 100mg Medchemexpress, Prasugrel (hydrochloride) CAS:389574-19-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15284A100MG
Encompass_Preferred
Medchemexpress, HY-15284A 100mg Prasugrel (hydrochloride) CAS:389574-19-0 Prasugrel hydrochloride (PCR 4099 hydrochloride), a thienopyridine and prodrug, inhibits platelet function. Prasugrel hydrochloride is an orally active and potent P2Y12 receptor antagonist, and inhibits ADP-induced platelet aggregation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1697