Learn More
Apexbio Technology LLC Prasugrel hydrochloride 389574-19-0 5mg
SDP
Catalog No. 5000087757
Encompass_Preferred
Prasugrel hydrochloride (CAS 389574-19-0) is a small molecule inhibitor that targets platelet activation by antagonizing the P2Y12 adenosine diphosphate (ADP) receptor on platelet surfaces By inhibiting this receptor prasugrel impedes ADP-mediated platelet aggregation a critical process in thrombus formation In biochemical assays prasugrel hydrochloride demonstrates an IC50 of 1 8 M for platelet inhibition This compound is utilized in biomedical research to investigate platelet signaling pathways and to model antithrombotic interventions in vitro
Catalog No. 50-000-87757 Supplier Apexbio Technology LLC Supplier No. B12835
Prasugrel hydrochloride (CAS 389574-19-0) is a small molecule inhibitor that targets platelet activation by antagonizing the P2Y12 adenosine diphosphate (ADP) receptor on platelet surfaces By inhibiting this receptor prasugrel impedes ADP-mediated platelet aggregation a critical process in thrombus formation In biochemical assays prasugrel hydrochloride demonstrates an IC50 of 1 8 M for platelet inhibition This compound is utilized in biomedical research to investigate platelet signaling pathways and to model antithrombotic interventions in vitro