Learn More
Medchemexpress LLC Prasugrel hydrochloride | 389574-19-0 | C20H21ClFNO3S | 1 ML
SDP
Catalog No. 5000372133
Encompass_Preferred
Prasugrel hydrochloride is a thienopyridine and proagent that inhibits platelet function. It is an orally active and potent P2Y12 receptor antagonist, inhibiting ADP-induced platelet aggregation. As an inactive prodrug, it requires metabolic processing to generate its active antiplatelet metabolite, which is rapidly absorbed from the gut.
  • Thienopyridine and proagent
  • Inhibits platelet function
  • Orally active and potent P2Y12 receptor antagonist
  • Inhibits ADP-induced platelet aggregation
  • Acts faster and is significantly more potent than clopidogrel in vivo
  • Requires metabolic processing to generate active antiplatelet metabolite
  • Rapidly absorbed from the gut
  • Maximum plasma levels of active metabolite achieved within 1 hour
  • Effective, maximum inhibition of platelet aggregation at 1-2 hours
Catalog No. 50-003-72133 Supplier Medchemexpress LLC Supplier No. HY15284A10MM/1ML
Prasugrel hydrochloride is a thienopyridine and proagent that inhibits platelet function. It is an orally active and potent P2Y12 receptor antagonist, inhibiting ADP-induced platelet aggregation. As an inactive prodrug, it requires metabolic processing to generate its active antiplatelet metabolite, which is rapidly absorbed from the gut.
  • Thienopyridine and proagent
  • Inhibits platelet function
  • Orally active and potent P2Y12 receptor antagonist
  • Inhibits ADP-induced platelet aggregation
  • Acts faster and is significantly more potent than clopidogrel in vivo
  • Requires metabolic processing to generate active antiplatelet metabolite
  • Rapidly absorbed from the gut
  • Maximum plasma levels of active metabolite achieved within 1 hour
  • Effective, maximum inhibition of platelet aggregation at 1-2 hours