Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pratensein | 2284-31-3 | 1 MG
SDP

Catalog No. 5000417359
Encompass_Preferred

Pratensein, a flavonoid, ameliorates β-amyloid-induced cognitive impairment in rats by reducing oxidative damage and restoring synapse and brain-derived neurotrophic factor (BDNF) levels. It attenuates neuronal degeneration and apoptosis in the hippocampus, and reduces IL-1β, TNF-α overexpression, astrogliosis, and microgliosis induced by Aβ1-42. Pratensein suppresses NF-κB activation and improves spatial learning and memory in Alzheimer's disease rat models.

  • Ameliorates β-amyloid-induced cognitive impairment.
  • Reduces oxidative damage; restores synapse and BDNF levels.
  • Attenuates neuronal degeneration and apoptosis.
  • Suppresses NF-κB activation in hippocampus.
  • Improves spatial learning and memory in Alzheimer's models.
  • Purity: 99.92%.
  • Appearance: solid, white to off-white.
  • Storage: -20°C, sealed.
  • For research use only.

Catalog No. 50-004-17359 Supplier Medchemexpress LLC Supplier No. HYN79811MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pratensein, a flavonoid, ameliorates β-amyloid-induced cognitive impairment in rats by reducing oxidative damage and restoring synapse and brain-derived neurotrophic factor (BDNF) levels. It attenuates neuronal degeneration and apoptosis in the hippocampus, and reduces IL-1β, TNF-α overexpression, astrogliosis, and microgliosis induced by Aβ1-42. Pratensein suppresses NF-κB activation and improves spatial learning and memory in Alzheimer's disease rat models.

  • Ameliorates β-amyloid-induced cognitive impairment.
  • Reduces oxidative damage; restores synapse and BDNF levels.
  • Attenuates neuronal degeneration and apoptosis.
  • Suppresses NF-κB activation in hippocampus.
  • Improves spatial learning and memory in Alzheimer's models.
  • Purity: 99.92%.
  • Appearance: solid, white to off-white.
  • Storage: -20°C, sealed.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.