Learn More
Cayman Chemical Pravastatin lactone, Purity: ≥98%, CAS: 85956-22-5, 10 mg
SDP
Catalog No. 502986899
Encompass_Preferred
A metabolite of pravastatin; formed when pravastatin undergoes acid catalyzed non-enzymatic lactonization in the stomach
Catalog No. 50-298-6899 Supplier Cayman Chemical Supplier No. 2257210 MG
A metabolite of pravastatin; formed when pravastatin undergoes acid catalyzed non-enzymatic lactonization in the stomach