Learn More
Medchemexpress LLC Pravastatin lactone | 85956-22-5 | 99.2% | 5 MG
SDP
Catalog No. 5000231541
Encompass_Preferred
Pravastatin lactone is the delta-lactone metabolite of pravastatin and a potent inhibitor of HMG-CoA reductase. It is used as a research reagent to study statin metabolism and cholesterol biosynthesis in biochemical and pharmacological assays.
  • Potent HMG-CoA reductase inhibitor used in cholesterol synthesis studies.
  • High purity suitable for analytical and biochemical applications.
  • Available in small milligram quantities for assay development and metabolism studies.
  • Molecular formula C23H34O6 and molecular weight 406.51 g/mol aid identification and dosing.
  • Provided as a neat solid for laboratory research use.
Catalog No. 50-002-31541 Supplier Medchemexpress LLC Supplier No. HY1217955MG
Pravastatin lactone is the delta-lactone metabolite of pravastatin and a potent inhibitor of HMG-CoA reductase. It is used as a research reagent to study statin metabolism and cholesterol biosynthesis in biochemical and pharmacological assays.
  • Potent HMG-CoA reductase inhibitor used in cholesterol synthesis studies.
  • High purity suitable for analytical and biochemical applications.
  • Available in small milligram quantities for assay development and metabolism studies.
  • Molecular formula C23H34O6 and molecular weight 406.51 g/mol aid identification and dosing.
  • Provided as a neat solid for laboratory research use.