Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pravastatin (sodium) | 81131-70-6 | 98.96% | 50 G
SDP

Catalog No. 5000406243
Encompass_Preferred

Pravastatin sodium (CS-514 sodium) is an HMG-CoA reductase inhibitor that targets sterol synthesis. It belongs to the statin drug class and is commonly used with diet and exercise to lower cholesterol and prevent cardiovascular disease. This product is supplied as an off-white to pale purple solid.

  • HMG-CoA reductase inhibitor against sterol synthesis
  • Used for lowering cholesterol and preventing cardiovascular disease
  • Soluble in DMSO and water
  • Appearance: off-white to pale purple solid

Catalog No. 50-004-06243 Supplier Medchemexpress LLC Supplier No. HYB0165A50G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Pravastatin sodium (CS-514 sodium) is an HMG-CoA reductase inhibitor that targets sterol synthesis. It belongs to the statin drug class and is commonly used with diet and exercise to lower cholesterol and prevent cardiovascular disease. This product is supplied as an off-white to pale purple solid.

  • HMG-CoA reductase inhibitor against sterol synthesis
  • Used for lowering cholesterol and preventing cardiovascular disease
  • Soluble in DMSO and water
  • Appearance: off-white to pale purple solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.