Learn More
Medchemexpress LLC Pravastatin (sodium) | 81131-70-6 | 99.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0165A5G
Encompass_Preferred
Pravastatin sodium is an HMG-CoA reductase inhibitor against sterol synthesis with an IC50 of 5.6 μM. It is a statin used in combination with diet, exercise, and weight-loss for lowering cholesterol and preventing cardiovascular disease.
  • Inhibits HMG-CoA reductase.
  • For research use only.
  • IC50 for inhibiting cellular steroidogenesis in HepG2 hepatocytes is 1.37 μM.
  • IC50 for antimalarial activity against Plasmodium berghei NK65 in HepG2 hepatocytes is > 200 μM (after 48 hours by RT-PCR).
  • Inhibits cholesterol synthesis in isolated rat hepatocytes with an IC50 of 0.1 μM.
  • Inhibits cholesterol synthesis from [14C]- Acetate in hepatocytes of rats with an IC50 of 100 nM.
Catalog No. 50-004-06295