Learn More
Medchemexpress LLC Pravastatin sodium | 81131-70-6 | 99.4% | 25 G
SDP
Catalog No. 5000407059
Encompass_Preferred
Pravastatin sodium is an HMG-CoA reductase inhibitor that targets sterol synthesis with an IC50 of 5.6 μM. It belongs to the drug class of statins, which are used to lower cholesterol and prevent cardiovascular disease, often in conjunction with diet, exercise, and weight loss.
  • Effectively inhibits sterol synthesis
  • Intended for research purposes
  • High purity
  • Soluble in DMSO (100 mg/mL) and H2O (≥ 50 mg/mL)
  • Used in cellular steroidogenesis inhibition studies in HepG2 cells
  • Cited in various publications for cell proliferation/viability assays, RT-PCR, and cell imaging/staining
Catalog No. 50-004-07059 Supplier Medchemexpress LLC Supplier No. 25GHYB0165A
Pravastatin sodium is an HMG-CoA reductase inhibitor that targets sterol synthesis with an IC50 of 5.6 μM. It belongs to the drug class of statins, which are used to lower cholesterol and prevent cardiovascular disease, often in conjunction with diet, exercise, and weight loss.
  • Effectively inhibits sterol synthesis
  • Intended for research purposes
  • High purity
  • Soluble in DMSO (100 mg/mL) and H2O (≥ 50 mg/mL)
  • Used in cellular steroidogenesis inhibition studies in HepG2 cells
  • Cited in various publications for cell proliferation/viability assays, RT-PCR, and cell imaging/staining