Learn More
Cayman Chemical Pravastatin (sodium salt), Purity: ≥98%, CAS: 81131-70-6, 50 mg
SDP
Catalog No. 502888645
Encompass_Preferred
A potent HMG-CoA reductase inhibitor (Ki = 2.3 nM); reduces total plasma cholesterol levels by 29% in dogs at 20 mg/kg per day for five weeks
Catalog No. 50-288-8645 Supplier Cayman Chemical Supplier No. 1001034350 MG
A potent HMG-CoA reductase inhibitor (Ki = 2.3 nM); reduces total plasma cholesterol levels by 29% in dogs at 20 mg/kg per day for five weeks