Learn More
Perkin Elmer US LLC Praying Mantis Accessory
Catalog No. 502112890
Encompass_Preferred
This invaluable diffuse reflection system includes: A cart with two mounted alignment mirrors A diffuse reflectance sampling cup - 10 mm in diameter with a volume of 0.25 mL An adjustable Microsampling cup - 3 mm in diameter with a volume of 0.03 ml) A Microsampling funnel Connection hardware
Catalog No. 50-211-2890 Supplier Perkin Elmer US LLC Supplier No. L6310211
Add to Cart
Add to Cart
Request a Quote
This invaluable diffuse reflection system includes: A cart with two mounted alignment mirrors A diffuse reflectance sampling cup - 10 mm in diameter with a volume of 0.25 mL An adjustable Microsampling cup - 3 mm in diameter with a volume of 0.03 ml) A Microsampling funnel Connection hardware