Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Praziquantel | 55268-74-1 | 99.7% | 25 G
SDP

Catalog No. 5000405599
Encompass_Preferred

Praziquantel is a broadly effective trematocide and cestocide. Its initial effect on the schistosome may involve an interaction with lipid constituents of the tegumental membrane. It is used for the research of schistosomiasis.

  • Broadly effective trematocide and cestocide
  • Used for the research of schistosomiasis
  • Appears as a white to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Soluble in DMSO (50 mg/mL)
  • Insoluble in water (< 0.1 mg/mL)
  • Enhances tension produced by musculature of male *Schistosomajaponicum*

Catalog No. 50-004-05599 Supplier Medchemexpress LLC Supplier No. 25GHYB0244
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Praziquantel is a broadly effective trematocide and cestocide. Its initial effect on the schistosome may involve an interaction with lipid constituents of the tegumental membrane. It is used for the research of schistosomiasis.

  • Broadly effective trematocide and cestocide
  • Used for the research of schistosomiasis
  • Appears as a white to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Soluble in DMSO (50 mg/mL)
  • Insoluble in water (< 0.1 mg/mL)
  • Enhances tension produced by musculature of male *Schistosomajaponicum*

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.