Learn More
Enzo Life Sciences Praziquantel (5g). CAS: 55268-74-1
SDP
Supplier:  Enzo Life Sciences ALX550321G005
Encompass_Preferred
Alternative name: 2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in 100% ethanol or chloroform; slightly soluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-9533