Learn More
Medchemexpress LLC Prazosin-d8 | 1006717-55-0 | 98.0% | 1 MG
SDP
Catalog No. 5000406975
Encompass_Preferred
Prazosin-d8 is a deuterium-labeled analog of Prazosin. It acts as an alpha-adrenergic blocker and is a sympatholytic drug primarily used for the treatment of high blood pressure, anxiety, PTSD, and panic disorder.
  • Deuterium labeled Prazosin
  • Functions as an alpha-adrenergic blocker
  • Sympatholytic drug
  • Used to treat high blood pressure, anxiety, PTSD, and panic disorder
  • Can be utilized as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may influence pharmacokinetic and metabolic profiles
Catalog No. 50-004-06975 Supplier Medchemexpress LLC Supplier No. HYB0193S1MG
Prazosin-d8 is a deuterium-labeled analog of Prazosin. It acts as an alpha-adrenergic blocker and is a sympatholytic drug primarily used for the treatment of high blood pressure, anxiety, PTSD, and panic disorder.
  • Deuterium labeled Prazosin
  • Functions as an alpha-adrenergic blocker
  • Sympatholytic drug
  • Used to treat high blood pressure, anxiety, PTSD, and panic disorder
  • Can be utilized as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may influence pharmacokinetic and metabolic profiles