Learn More
Medchemexpress LLC Prazosin-d8 | 1006717-55-0 | 98.0% | 5 MG
SDP
Catalog No. 5000405789
Encompass_Preferred
Prazosin-d8 is a deuterium labeled form of Prazosin. Prazosin is an alpha-adrenergic blocker and sympatholytic drug. It is used to treat conditions such as high blood pressure, anxiety, PTSD, and panic disorder.
  • Deuterium labeled compound
  • Alpha-adrenergic blocker
  • Sympatholytic drug
  • Used in high blood pressure treatment
  • Applied in managing anxiety and PTSD
  • Effective in treating panic disorder
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05789 Supplier Medchemexpress LLC Supplier No. HYB0193S5MG
Prazosin-d8 is a deuterium labeled form of Prazosin. Prazosin is an alpha-adrenergic blocker and sympatholytic drug. It is used to treat conditions such as high blood pressure, anxiety, PTSD, and panic disorder.
  • Deuterium labeled compound
  • Alpha-adrenergic blocker
  • Sympatholytic drug
  • Used in high blood pressure treatment
  • Applied in managing anxiety and PTSD
  • Effective in treating panic disorder
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs