Learn More
Medchemexpress LLC Prazosin-d8 Hydrochloride | 1189508-34-6 | 99.8% | 5 MG
SDP
Catalog No. 5000406173
Encompass_Preferred
Prazosin-d8 hydrochloride is the deuterium labeled Prazosin hydrochloride. Prazosin hydrochloride is an alpha-adrenergic blocker.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06173 Supplier Medchemexpress LLC Supplier No. HYB0193AS5MG
Prazosin-d8 hydrochloride is the deuterium labeled Prazosin hydrochloride. Prazosin hydrochloride is an alpha-adrenergic blocker.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.