Learn More
Apexbio Technology LLC Prazosin HCl,100mg CAS# 19237-84-4
SDP
Catalog No. 502240581
Encompass_Preferred
α1 and α2B-adrenoceptor antagonist. Also a potent antagonist at the melatonin MT3 receptor (Ki = 10.2 nM). Also available as part of the α1-Adrenoceptor Tocriset™ and Mixed Adre. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0581 Supplier Apexbio Technology LLC Supplier No. A5318100
α1 and α2B-adrenoceptor antagonist. Also a potent antagonist at the melatonin MT3 receptor (Ki = 10.2 nM). Also available as part of the α1-Adrenoceptor Tocriset™ and Mixed Adre. Other sizes are also available. Please inqury us for quote.