Learn More
Medchemexpress LLC Prazosin (hydrochloride) | 19237-84-4 | 99.9% | 200 MG
SDP
Catalog No. 5000406621
Encompass_Preferred
Prazosin hydrochloride is a well-tolerated, CNS-active α1-adrenergic receptor antagonist. It is utilized in research for high blood pressure and alcohol use disorders. The compound effectively inhibits Norepinephrine (NE)-stimulated 45Ca efflux with an IC50 of 0.15 nM and also inhibits organic cation transporters OCT-1 and OCT-3 with IC50s of 1.8 and 13 μM, respectively.
  • Acts as a CNS-active α1-adrenergic receptor antagonist
  • Used in research for high blood pressure
  • Used in research for alcohol use disorders
  • Potently inhibits Norepinephrine (NE)-stimulated 45Ca efflux
  • Inhibits organic cation transporters OCT-1 and OCT-3
Catalog No. 50-004-06621 Supplier Medchemexpress LLC Supplier No. HYB0193A200MG
Prazosin hydrochloride is a well-tolerated, CNS-active α1-adrenergic receptor antagonist. It is utilized in research for high blood pressure and alcohol use disorders. The compound effectively inhibits Norepinephrine (NE)-stimulated 45Ca efflux with an IC50 of 0.15 nM and also inhibits organic cation transporters OCT-1 and OCT-3 with IC50s of 1.8 and 13 μM, respectively.
  • Acts as a CNS-active α1-adrenergic receptor antagonist
  • Used in research for high blood pressure
  • Used in research for alcohol use disorders
  • Potently inhibits Norepinephrine (NE)-stimulated 45Ca efflux
  • Inhibits organic cation transporters OCT-1 and OCT-3