Learn More
Sigma Aldrich Fine Chemicals Biosciences Prazosin hydrochloride 99.50MG
Catalog No. 502899741
Encompass
Prazosin is a member of the class of adrenergic blockers. It is considered as a peripheral vasodilator antihypertensive agent. Prazosin is a derivative of quinazoline.
Catalog No. 50-289-9741 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P779150MG
Prazosin is a member of the class of adrenergic blockers. It is considered as a peripheral vasodilator antihypertensive agent. Prazosin is a derivative of quinazoline.