Learn More
Sigma Aldrich Fine Chemicals Biosciences Prazosin hydrochloride Uni50MG
Catalog No. 502902912
Encompass
Prazosin hydrochloride is an alpha-adrenergic blocking agent employed as an antihypertensive drug for the treatment of high blood pressure. It lowers the blood pressure by binding to the alpha receptors thereby relaxing the blood vessels.
Catalog No. 50-290-2912 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1554705350MG
Prazosin hydrochloride is an alpha-adrenergic blocking agent employed as an antihypertensive drug for the treatment of high blood pressure. It lowers the blood pressure by binding to the alpha receptors thereby relaxing the blood vessels.