Learn More
Aviva Systems Biology Corp PRDM12 Antibody - N-terminal region (ARP33218_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33218_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDM12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRDM12. Alternate names: PFM9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-54921