Learn More
Aviva Systems Biology Corp Prdm12 Antibody - middle region (ARP57895_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57895_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prdm12. It was validated on Western Blot. . Alternate names: Gm998ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-410