Learn More
Aviva Systems Biology Corp PRDM13 Antibody - C-terminal region (ARP39506_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39506_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDM13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PRDM13. Alternate names: PFM10, MU-MB-20.220ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57063