Learn More
Aviva Systems Biology Corp PRDM15 Antibody - C-terminal region (ARP36148_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDM15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PRDM15. Alternate names: C21orf83, PFM15, ZNF298ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-56031