Learn More
Aviva Systems Biology Corp PRDM9 Antibody - middle region (ARP35967_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35967_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDM9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRDM9. Alternate names: PFM6, MSBP3, PRMD9, ZNF899, MEISETZApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-55948