Learn More
Aviva Systems Biology Corp PRDX1 Antibody - N-terminal region (ARP48454_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48454_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRDX1. Alternate names: MSP23, NKEFA, PAG, PAGA, PAGB, PRX1, TDPX2, PRXIApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59833