Learn More
Aviva Systems Biology Corp PRDX4 Antibody - C-terminal region (ARP64307_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64307_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDX4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human PRDX4. Alternate names: AOE37-2, PRX-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-232