Learn More
Aviva Systems Biology Corp PRDX5 Antibody - middle region (ARP54832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDX5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRDX5. Alternate names: ACR1, AOEB166, B166, MGC117264, MGC142283, MGC142285, PLP, PMP20, PRDX6, PRXV, SBBI10ApplicationWB. Species reactivity: Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-61334