Learn More
Aviva Systems Biology Corp PRDX6 Antibody - C-terminal region (ARP48268_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48268_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRDX6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PRDX6. Alternate names: 1-Cys, AOP2, KIAA0106, MGC46173, NSGPx, PRX, aiPLA2, p29ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59777