Learn More
Selleck Chemical LLC PRE-084 hydrochloride
SDP
Catalog No. 5000002476
Encompass_Preferred
PRE-084 hydrochloride (HCl) is a potent selective sigma-1 receptor ( 1 S1R) agonist with IC50 of 44 nM in the sigma receptor assay
Catalog No. 50-000-02476 Supplier Selleck Chemical LLC Supplier No. S69505MG
PRE-084 hydrochloride (HCl) is a potent selective sigma-1 receptor ( 1 S1R) agonist with IC50 of 44 nM in the sigma receptor assay