Learn More
Selleck Chemical LLC PRE-084 hydrochloride
Supplier: Selleck Chemical LLC S695025MG
PRE-084 hydrochloride (HCl) is a potent selective sigma-1 receptor ( 1 S1R) agonist with IC50 of 44 nM in the sigma receptor assay