Learn More
Cayman Chemical PRE-084hydrochlorIde 50mg
SDP
Catalog No. 502893738
Encompass_Preferred
A selective sigma σ1 receptor agonist (IC50 = 44 nM; Kis = 2.2 and 13,091 nM for σ1 and σ2 receptors, respectively) with nootropic and antidepressant actions in vivo, as well as antitussive and reinforcing effects
Catalog No. 50-289-3738 Supplier Cayman Chemical Supplier No. 900132950 MG
A selective sigma σ1 receptor agonist (IC50 = 44 nM; Kis = 2.2 and 13,091 nM for σ1 and σ2 receptors, respectively) with nootropic and antidepressant actions in vivo, as well as antitussive and reinforcing effects