Learn More
MilliporeSigma™ Supelco™ Pre-Activated Florisil™, Ampulized
Catalog No. 111010815 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 Pk.
Catalog No. Quantity
11-101-0815 10 Pk.
1 options
Catalog No. 11-101-0815 Supplier MilliporeSigma™ Supelco™ Supplier No. 48924U
Add to Cart
Add to Cart
Intended for use with the Supelco™ Dioxin Prep System to affect the class separation of polychlorinated biphenyls (PCBs) from polychlorinated dibenzo-p-dioxins and dibenzo-furans (PCDD/Fs).
  • Particle Size: 60/100 Mesh
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 Pk.
For Use With (Equipment) Supelco™ Dioxin Prep System
Florisil is a registered trademark of U.S. Silica Company