Learn More
Zen Bio Inc PRE ADIPOCYTE MEDIA 500ML
SDP
Catalog No. NC9860640
Encompass
Cell Culture Media; Preadipocyte Media; Zen-Bio; For plating preadipocytes prior to differentiation; Contains growth factors needed for expansion of subcutaneous preadipocytes; Ready-to-use; 500mL
Catalog No. NC9860640 Supplier Zen Bio Inc Supplier No. PM1
Cell Culture Media; Preadipocyte Media; Zen-Bio; For plating preadipocytes prior to differentiation; Contains growth factors needed for expansion of subcutaneous preadipocytes; Ready-to-use; 500mL