Learn More
Grainger Pre Amplifier Ryton PPS 8 Pin DIN
Catalog No. 502767640
Encompass_Preferred
The Grainger Ryton PPS electrodes and probes are made of durable Ryton polyphenylene sulfide plastic for chemical resistance. They are ideal for water quality testing and monitoring pH, ORP, conductivity, and other parameters. The selection includes various styles like spear tip, flat surface, and bulb tip to suit different applications. Key features are quick response times, accuracy, and compatibility with meters. These probes stand up to acids, bases, and solvents for reliable measurements in harsh environments. Grainger offers a range of sizes, connector types, cable lengths, and price points to meet diverse needs.
Catalog No. 50-276-7640 Supplier Grainger Supplier No. 16Z033
The Grainger Ryton PPS electrodes and probes are made of durable Ryton polyphenylene sulfide plastic for chemical resistance. They are ideal for water quality testing and monitoring pH, ORP, conductivity, and other parameters. The selection includes various styles like spear tip, flat surface, and bulb tip to suit different applications. Key features are quick response times, accuracy, and compatibility with meters. These probes stand up to acids, bases, and solvents for reliable measurements in harsh environments. Grainger offers a range of sizes, connector types, cable lengths, and price points to meet diverse needs.