Learn More
Analytical Sales & Services Pre-Assembed 13mm Black PP Screw Caps with Tan Silicone/PTFE Liners
Catalog No. 5000804539
Encompass
Advantage™ Pre-Assembed 13mm Black Polypropylene Screw Caps with Tan Silicone/PTFE Liners
Catalog No. 50-008-04539 Supplier Analytical Sales & Services Supplier No. 53009CASE
Advantage™ Pre-Assembed 13mm Black Polypropylene Screw Caps with Tan Silicone/PTFE Liners