Learn More
Waters Corp WATERS CORPORATION Vial, Screw Neck, 12 x 32 mm, Clear Glass, Pre-Assembled
Catalog No. 50780523 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These clear glass screw neck vials are pre-assembled with a pre-slit septum for dissolution systems. They have a 2 mL volume capacity and are designed to withstand high temperatures during sample preparation. Compatible with most common autosamplers, these vials provide a secure seal with a pre-slit PTFE cap and are suitable for both liquid and solid sample storage.
  • Pre-assembled vial with pre-slit septum for dissolution systems
  • Screw neck closure, 12 x 32 mm dimensions
  • Clear glass material with 2 mL volume capacity
  • Autosampler compatible, ideal for sample preparation
  • Includes vial, cap, and septum
  • Withstands high temperatures for sample processing
  • Pre-slit PTFE cap for secure sealing
  • Suitable for liquid and solid sample storage
  • Packaged in units of 100
Catalog No. 50-780-523 Supplier Waters Corp Supplier No. 186000989
These clear glass screw neck vials are pre-assembled with a pre-slit septum for dissolution systems. They have a 2 mL volume capacity and are designed to withstand high temperatures during sample preparation. Compatible with most common autosamplers, these vials provide a secure seal with a pre-slit PTFE cap and are suitable for both liquid and solid sample storage.
  • Pre-assembled vial with pre-slit septum for dissolution systems
  • Screw neck closure, 12 x 32 mm dimensions
  • Clear glass material with 2 mL volume capacity
  • Autosampler compatible, ideal for sample preparation
  • Includes vial, cap, and septum
  • Withstands high temperatures for sample processing
  • Pre-slit PTFE cap for secure sealing
  • Suitable for liquid and solid sample storage
  • Packaged in units of 100