Learn More
Analytical Sales & Services Pre-Assembled 13mm Black PP Screw Caps with Pre-slit 0.65" Red Silicone/PTFE Liners
Catalog No. 5000804320
Encompass
Advantage™ Pre-Assembled 13mm Polypropylene Screw Caps with 0.65" Red Silicone/PTFE Liners
Catalog No. 50-008-04320 Supplier Analytical Sales & Services Supplier No. 53126SLCASE
Advantage™ Pre-Assembled 13mm Polypropylene Screw Caps with 0.65" Red Silicone/PTFE Liners