Learn More
Analytical Sales & Services 9mm, 2ml Clear Wide Mouth Twist Vials And Blue Caps with PTFE/Silicone Liners
Catalog No. 50450664
Encompass
This item has a minimum order quantity of 2 per supplier requirements. PRE-ASSEMBLED 2ML CLEAR 100PK
Catalog No. 50-450-664 Supplier Analytical Sales & Services Supplier No. 58128
This item has a minimum order quantity of 2 per supplier requirements. PRE-ASSEMBLED 2ML CLEAR 100PK