Learn More
AB Sciex LLC Pre-Assembled Capillary Cartridge
Catalog No. NC1107665
Encompass
CE-SDS can be performed using factory manufactured cartridges, pre-assembled with bare fused-silica capillaries. Pre-assembled cartridges ensure precision cut ends and alignment of capillary windows with the detection path resulting in maximized consistency of your separations.
Catalog No. NC1107665 Supplier AB Sciex LLC Supplier No. A55625
CE-SDS can be performed using factory manufactured cartridges, pre-assembled with bare fused-silica capillaries. Pre-assembled cartridges ensure precision cut ends and alignment of capillary windows with the detection path resulting in maximized consistency of your separations.