Learn More
Premier Lab Supply CEMBLES Pre-Assembled XRF Sample Cups – PN# PA-420-45-135
SDP
Catalog No. NC1287876
Encompass_Preferred
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-420-45-135; 45mm Cups PN# SC-3345; Mylar film 3.5μm; 32 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.
Catalog No. NC1287876 Supplier Premier Lab Supply Supplier No. PA42045135
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-420-45-135; 45mm Cups PN# SC-3345; Mylar film 3.5μm; 32 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.