Learn More
Chemplex Industries PRE-ASSEMEMBLED SAMPLE CUP
Catalog No. NC2313663
Encompass
Stackable Analyzer TrimLess XRF Sample Cups Single Open End 1.69 42.9mm Dia X 0.73 18.5mm Height with 2.5um Mylar 22 sets/tube - Designed to fit Horiba 6100/6800/60A Series
Catalog No. NC2313663 Supplier Chemplex Industries Supplier No. 10P8713
Stackable Analyzer TrimLess XRF Sample Cups Single Open End 1.69 42.9mm Dia X 0.73 18.5mm Height with 2.5um Mylar 22 sets/tube - Designed to fit Horiba 6100/6800/60A Series