Learn More
Bio Plas Pre-barcoded PermaFrame 384-Well Full-Skirt PCR Plate with A24 and P24 Notch and Clear Wells
Catalog No. 22038958
Change view
Click to view available options
Color:
Clear
Catalog No. Color
22-038-958 Clear
1 options
Catalog No. 22-038-958 Supplier Bio Plas Supplier No. 9000BC
Add to Cart
Add to Cart
384-well PCR plate with full skirt.
  • PermaFrame 384-well full-skirt PCR plate
  • A24/P24 notches
  • Clear wells
  • Pre-barcoded
  • Clear
Specifications
Barcode Includes Barcode
Format Standard
Array 16 x 24
Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
Product Type PCR Plate
Material Polycarbonate
Volume (Metric) Well 30 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
Fit for Purpose PCR Ready