Learn More
Bio Plas Pre-barcoded PermaFrame 96-Well Full-Skirt PCR Plate with White Wells
Catalog No. 22038964
Change view
Click to view available options
Color:
White, Clear
Catalog No. Color
22-038-964 White, Clear
1 options
Catalog No. 22-038-964 Supplier Bio Plas Supplier No. 9200WBC
Add to Cart
Add to Cart
96-well PCR plate with full skirt.
  • PermaFrame 96-Well full-skirt 0.1 mL PCR plate
  • White wells
  • Pre-barcoded
  • Clear and white
Specifications
Barcode Includes Barcode
Format Standard
Array 8 x 12
Autoclavable Not Autoclavable
Color White, Clear
Disposable Single-use
Product Type PCR Plate
Material Polycarbonate
Volume (Metric) Well 0.1 mL
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
RNase Free Yes
Fit for Purpose PCR Ready